Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP12 Mouse anti-Human, DyLight™ 550, Clone: OTI1E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274801R

Catalog No. NBP274801R


Add to cart

Description

Description

USP12 Monoclonal antibody specifically detects USP12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

USP12
OTI1E3
Western Blot
Affinity Purified, Conjugated
USP12
Recombinant protein fragment corresponding to amino acids 61-251 of human USP12 (NP_872294) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Deubiquitinating enzyme 12, EC 3.1.2.15, EC 3.4.19.12, UBH1ubiquitin specific protease 12, ubiquitin carboxyl-terminal hydrolase 12, ubiquitin specific peptidase 12, ubiquitin specific protease 12 like 1, ubiquitin thioesterase 12, Ubiquitin thiolesterase 12, Ubiquitin-hydrolyzing enzyme 1, Ubiquitin-specific-processing protease 12, USP12L1
Mouse
IgG1
0.1 mL
Primary
219333.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit