Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP13 Mouse anti-Human, Canine, Monkey, HRP, Clone: OTI4G11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274802H

Catalog No. NBP274802H


Add to cart

Description

Description

USP13 Monoclonal antibody specifically detects USP13 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

USP13
OTI4G11
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
USP13
Full-length protein expressed in 293T cell transfected with human USP13 expression vector.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey
Immunohistochemistry, Western Blot
HRP
PBS with No Preservative
Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, ISOT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13
Mouse
IgG1
0.1 mL
Primary
8975.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit