Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP14, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300920AM

 View more versions of this product

Catalog No. 50-155-8794


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. Mice with a mutation that results in reduced expression of the ortholog of this protein are retarded for growth, develop severe tremors by 2 to 3 weeks of age followed by hindlimb paralysis and death by 6 to 10 weeks of age. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

USP14
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
P54578, Q9JMA1
Ubiquitin carboxyl-terminal hydrolase 14; Ubiquitin thiolesterase 14; Ubiquitin-specific processing protease 14; Deubiquitinating enzyme 14
Rabbit
IgG
100 μl
4° C
Primary
59025, 9097
Immunohistochemistry, Western Blot
Unconjugated
USP14
liquid
Usp14
Between 450 and C-term
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit