Learn More
Abnova™ USP15 Recombinant Protein
Human full-length ORF recombinant protein with GST-tag at N-terminal
$503.00 - $775.00
Specifications
Accession Number | AAH20688 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 9958 |
Molecular Weight (g/mol) | 52 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-011-700
|
Abnova™
H00009958P0110 |
10 ug |
Each for $503.00
|
|
|||||
89-011-701
|
Abnova™
H00009958P0125 |
25 ug |
Each for $775.00
|
|
|||||
Description
- Encoded by ubiquitin specific peptidase 15 gene
- Molecular weight: 51.59kDa
- Preparation method: in vitro wheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50 mM Tris-HCI, 10 mM reduced glutathione
- pH=8.0 in elution buffer
Sequence: MAEGGAADLDTQRSDIATLLKTSLRKGDTWYLVDSRWFKQWKKYVGFDSWDKYQMGDQNVYPGPIDNSGLLKDGDAQSLKEHLIDELDYILLPTEGWNKLVSWYTLMEGQEPIARKVVEQGMFVKHCKVEVYLTELKLCENGNMNNVVTRRFSKADTIDTIEKEIRKIFSIPDEKETRLWNKYMSNTF
EPLNKPDSTIQDAGLYQGQVLVIEQKNEDGTWPRGPSTPKKPLEQSC
Best when used within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH20688 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
52 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
USP15 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
9958 | |
USP15 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEGGAADLDTQRSDIATLLKTSLRKGDTWYLVDSRWFKQWKKYVGFDSWDKYQMGDQNVYPGPIDNSGLLKDGDAQSLKEHLIDELDYILLPTEGWNKLVSWYTLMEGQEPIARKVVEQGMFVKHCKVEVYLTELKLCENGNMNNVVTRRFSKADTIDTIEKEIRKIFSIPDEKETRLWNKYMSNTFEPLNKPDSTIQDAGLYQGQVLVIEQKNEDGTWPRGPSTPKKPLEQSC | |
KIAA0529/MGC131982/MGC149838/MGC74854/UNPH4 | |
USP15 | |
Wheat Germ (in vitro) | |
GST |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.