Learn More
Invitrogen™ USP20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533103
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Dog, Bat, Bovine, Elephant, Panda, Horse, Rabbit, Pig, Opossum, Turkey, Chicken, Platypus (100%); Mouse, Rat, Hamster (93%); Zebrafish, Stickleback (87%); Pufferfish (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications
USP20 | |
Polyclonal | |
Unconjugated | |
USP20 | |
1700055M05Rik; AI467231; Deubiquitinating enzyme 20; hVDU2; KIAA1003; LSFR3A; pVHL-interacting deubiquitinating enzyme 2; ubiquitin carboxyl-terminal hydrolase 20; ubiquitin specific peptidase 20; ubiquitin specific protease 20; ubiquitin thioesterase 20; ubiquitin thiolesterase 20; ubiquitin-specific-processing protease 20; Usp20; Vdu2; VHL-interacting deubiquitinating enzyme 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100066778, 100157102, 100341397, 10868, 417188, 480696, 505839 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 0.1% sodium azide | |
A7Z056, Q9Y2K6 | |
USP20 | |
Synthetic 15 amino acid peptide from internal region of human USP20. | |
50 μg | |
Primary | |
Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Chicken, Bat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.