Learn More
USP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 174911AP
Catalog No. 50-173-1602
Add to cart
Description
Description
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications
Specifications
USP20
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C6M1, Q9Y2K6
Deubiquitinating enzyme 20, hVDU2, KIAA1003, LSFR3A, Ubiquitin thiolesterase 20, USP20, VDU2
Rabbit
IgG
150 μL
-20°C
Primary
10868, 311856, 74270
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
USP20
Liquid
Usp20
USP20 Fusion Protein Ag11590
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat