Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP22 Rabbit anti-Human, Clone: JU63-27, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275702

Catalog No. NBP275702


Add to cart

Description

Description

USP22 Monoclonal antibody specifically detects USP22 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

USP22
JU63-27
Unconjugated
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L
Rabbit
IgG
Protein A purified
Cancer, Cell Cycle and Replication, Signal Transduction, Stem Cell Markers
Primary
23326.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Purified
USP22
Recombinant Protein
60 kDa
100 μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit