Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP22 Rabbit anti-Human, Mouse, DyLight™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149644UV

Catalog No. NBP149644UV


Add to cart

Description

Description

USP22 Polyclonal antibody specifically detects USP22 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

USP22
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
USP22
A genomic peptide made to an internal region of the human USP22 protein (within residues 100-205). [Swiss-Prot Q9H3R0]
Immunogen affinity purified
Cancer, Cell Cycle and Replication, Signal Transduction, Stem Cell Markers
Primary
23326.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit