Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Usp27x Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570389
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 85%; Zebrafish: 100%.
Deubiquitinase that can reduce the levels of BCL2L11/BIM ubiquitination and stabilize BCL2L11 in response to the RAF-MAPK-degradation signal. By acting on BCL2L11 levels, may counteract the anti-apoptotic effects of MAPK activity.
Specifications
Usp27x | |
Polyclonal | |
Unconjugated | |
USP27X | |
Deubiquitinating enzyme 27; RGD1564628; Sfc11; Ubiquitin carboxyl-terminal hydrolase 22-like; Ubiquitin carboxyl-terminal hydrolase 27; ubiquitin carboxyl-terminal hydrolase 51-like; ubiquitin specific peptidase 27 X-linked; ubiquitin specific peptidase 27, X chromosome; ubiquitin specific peptidase 27, X-linked; ubiquitin specific peptidase 27, X-linked, pseudogene 1; ubiquitin specific protease 27, X chromosome; ubiquitin thioesterase 27; ubiquitin thiolesterase 22; ubiquitin thiolesterase 27; ubiquitin-specific-processing protease 27; USP22L; USP27; Usp27x; Usp27x-ps1; X-linked ubiquitin carboxyl-terminal hydrolase 27; X-linked ubiquitin thioesterase 27 | |
Rabbit | |
Affinity chromatography | |
RUO | |
54651 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8CEG8 | |
USP27X | |
synthetic peptide directed towards the middle region of human Usp27x (aa 91-140). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction