Learn More
USP40 Mouse anti-Human, Clone: OTI7D1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809899
Catalog No. 50-166-9781
Part Not Found
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be catalytically inactive.
Specifications
Specifications
USP40
Monoclonal
Unconjugated
USP40
Lyophilized
USP40
Human recombit protein fragment corresponding to amino acids 767-912 of human USP40 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI7D1
PBS with 8% trehalose and no preservative; pH 7.3
Q9NVE5
ubiquitin specific peptidase 40
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55230