Learn More
Novus Biologicals™ USP40 Knockout HeLa Cell Lysate
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Manufacturer:  Novus Biologicals™ NBP265230
Catalog No. NBP265230
Add to cart
Description
Description
The USP40 Knockout HeLa Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
LYOPH
Human
2 Vials
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Western Blot
Knockout achieved by using CRISPR/Cas9,-2 bp deletion in exon1
0.1 mg cell homogenate lyophilized in RIPA buffer made with double-knockout cell lines
USP40