Learn More
USP42 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 188111AP
Catalog No. 50-173-2236
Add to cart
Description
Description
This antibody is specific to USP42.
USP42 (ubiquitin specific peptidase 42) is a 1,325 amino acid protein that belongs to the peptidase C19 family. Expressed in a variety of tissues, USP42 functions to catalyze the conversion of a ubiquitin C-terminal thioester to a free ubiquitin and a thiol. The gene encoding USP42 maps to human chromosome 7, which houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.
Specifications
Specifications
USP42
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B2RQC2, D3ZU96, Q9H9J4
USP42
Rabbit
IgG
150 μL
-20°C
Primary
288482, 76800, 84132
Western Blot
Unconjugated
USP42
Liquid
Usp42
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat