Learn More
USP45 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158661AP
Catalog No. 50-173-0540
Add to cart
Specifications
Specifications
USP45
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q70EL2, Q8K387
Deubiquitinating enzyme 45, Ubiquitin thiolesterase 45, USP45
Rabbit
IgG
150 μL
-20°C
Primary
313098, 77593, 85015
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
USP45
Liquid
USP45
USP45 Fusion Protein Ag8478
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat