Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP46 Mouse anti-Human, Clone: CL0364, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524601

Catalog No. PIMA524601


Add to cart

Description

Description

Immunogen sequence: LFDNYMQQDA HEFLNYLLNT IADILQEEKK QEKQNGKLKN GNMNEPAENN KPELTWVHEI FQGTLTNETR CLNCETVSSK DEDFLDLSVD VEQN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%. Binds to an epitope located within the peptide sequence TNETRCLNCE as determined by overlapping synthetic peptides.

Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes (Quesada et al., 2004 [PubMed 14715245]).
Specifications

Specifications

USP46
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P62068
deubiquitinating enzyme 46, ubiquitin carboxyl-terminal hydrolase 46, ubiquitin specific protease 46, ubiquitin thioesterase 46, ubiquitin thiolesterase 46, ubiquitin-specific-processing protease 46
Mouse
IgG2a
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
64854
Immunofluorescence, Western Blot
CL0364
Unconjugated
USP46
Liquid
USP46
Recombit protein corresponding to Human USP46
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only