Learn More
USP50 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 203741AP
Catalog No. 50-173-2855
Add to cart
Description
Description
The antibody is specific to USP50.
Has no peptidase activity.
Specifications
Specifications
USP50
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P8X6, Q70EL3
Inactive ubiquitin carboxyl-terminal hydrolase 50, USP50
Rabbit
IgG
150 μL
-20°C
Primary
311399, 373509, 75083
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
USP50
Liquid
Usp50
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat