Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP7 Mouse anti-Human, DyLight 405, Clone: 6512, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243619V

Catalog No. NBP243619V


Add to cart

Description

Description

USP7 Monoclonal antibody specifically detects USP7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

USP7
6512
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
USP7
Recombinant protein encompassing a sequence within the N-terminus region of human USP7. The exact sequence is proprietary.
Protein G purified
Apoptosis, Cell Biology, DNA Repair, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway
Primary
7874
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Deubiquitinating enzyme 7, EC 3.1.2.15, EC 3.4.19.12, HAUSPTEF1, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, ubiquitin carboxyl-terminal hydrolase 7, ubiquitin specific peptidase 7 (herpes virus-associated), ubiquitin specific protease 7 (herpes virus-associated), Ubiquitin thioesterase 7, Ubiquitin-specific-processing protease 7
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit