Learn More
MCR Safety USS Defense 2 Foam
Click to view available options
Lens Tint:
Clear
Gray
Lens Coating:
Anti-fog
Description
- Entire Series Meets Military Ballistic High Velocity Standard
- MAX6™ Premium Anti-Fog Coating
- Sleek Co-Injected TPR over polycarbonate temples
- Fixed Closed Cell Foam Lined option
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015, CSA Z94.3 and MIL - PRF - 31013 standards.
Specifications
Lens Tint | Clear |
For Use With (Application) | Manufacturing |
Frame Color | Black |
No. per Pack | 1 Pair |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |