Learn More
Sklar InstrumentsSupplier Diversity Partner Hegar Uterine Dilator, Sklar, Curved, Double End 13mm/14mm, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 904814
Hegar Uterine Dilator, Sklar, Curved, Double End 13mm/14mm, Stainless Steel. Sklar's Hegar Double-Ended Uterine Dilator is a manual, probe-style instrument used to dilate the cervix. They are solid, nonflexible, and available in multiple diameters. These dilators are slightly curved in opposite directions from the center and have smooth, rounded tips. This product is the 13mm/14mm dilator. The overall length is 8 inches.
Catalog No. 50-143-6741