Learn More
Sklar InstrumentsSupplier Diversity Partner Simpson Uterine Sound, Sklar, 12-1/2 in. Simpson Uterine Sound Centimeter Graduation Silver Plated. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 905413
Simpson Uterine Sound, Sklar, 12-1/2 in. Simpson Uterine Sound Centimeter Graduation Silver Plated. Sklar's Simpson Uterine Sound is a malleable, silver plated OB/GYN instrument used for probing and dilating the uterus, as well as measuring the length of the cervical canal. It is round and thin with a small nodule at the tip and a thumb grip handle. It is very similar to the Sims uterine sound. This product has a gently curved tip, graduated centimeter markings, and an overall length of 12-1/2 inches.
Catalog No. 50-143-6822