Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uteroglobin Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Uteroglobin |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6347
![]() ![]() |
Proteintech Group Inc
104901AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Uteroglobin encodes a member of the secretoglobin family of small secreted proteins. The encoded protein has been implicated in numerous functions including anti-inflammation, inhibition of phospholipase A2 and the sequestering of hydrophobic ligands. Defects in this gene are associated with a susceptibility to asthma.Specifications
Uteroglobin | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P11684, Q06318 | |
22287, 7356 | |
Uteroglobin Fusion Protein Ag0758 | |
Primary | |
-20°C | |
SCGB1A1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CC10, CC16, CCPBP, CCSP, SCGB1A1, UGB, UP 1, UP1, Urinary protein 1, Urine protein 1, Uteroglobin | |
SCGB1A1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title