Learn More
Uteroglobin/SCGB1A1 Mouse anti-Human, HRP, Clone: 3A8B8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237439H
Catalog No. NBP237439H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Uteroglobin/SCGB1A1
3A8B8
Western Blot, ELISA
Purified
SCGB1A1
Purified recombinant fragment of human SCGB1A1 (AA: 26-91) expressed in E. Coli.
Protein A or G purified
Biologically Active Proteins, Cytokine Research
Primary
7356
Western Blot, ELISA
HRP
PBS with No Preservative
blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human