Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uteroglobin/SCGB1A1 Rabbit anti-Human, Clone: 135, Novus Biologicals™
Rabbit Monoclonal Antibody
Specifications
Antigen | Uteroglobin/SCGB1A1 |
---|---|
Clone | 135 |
Dilution | Flow Cytometry 1:25-1:100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB020783
|
Novus Biologicals
NBP289650100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB020784
|
Novus Biologicals
NBP28965050UL |
50 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Uteroglobin/SCGB1A1 Monoclonal specifically detects Uteroglobin/SCGB1A1 in Human samples. It is validated for Flow Cytometry.Specifications
Uteroglobin/SCGB1A1 | |
Flow Cytometry 1:25-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
blastokinin, CC10Urine protein 1, CC16, CCSPCCPBP, Clara cell phospholipid-binding protein, Clara cells 10 kDa secretory protein, Secretoglobin family 1A member 1, secretoglobin, family 1A, member 1 (uteroglobin), UGBUP-1, UP1, Urinary protein 1, uteroglobin | |
Recombinant Human Uteroglobin/SCGB1A1 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
135 | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cytokine Research | |
0.2 um filtered solution in PBS | |
7356 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title