Learn More
UTF1 Mouse anti-Human, DyLight 650, Clone: 5B6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237444C
Catalog No. NBP237444C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
UTF1
5B6
Western Blot, Flow Cytometry, ELISA
Purified
UTF1
Purified recombinant fragment of human UTF1 (AA: 148-214) expressed in E. Coli.
Protein A or G purified
Stem Cell Markers
Primary
8433
ELISA, Flow Cytometry, Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
undifferentiated embryonic cell transcription factor 1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human