Learn More
Sklar InstrumentsSupplier Diversity Partner Utility Sterilization Forceps, Sklar, Curved, 11 in., Stainless Steel. This product has wide tips for easy grasping. It has an overall length of 11 inches. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101211
Utility Sterilization Forceps, Sklar, Curved, 11 in., Stainless Steel. Sklar Utility Sterilization Forceps are used for placement and removal of instruments and surgical equiptment into and out of a sterilizer. This product has wide tips for easy grasping. It has an overall length of 11 inches.
Catalog No. 50-143-6948