Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Multi-Cut Utility Scissors, Sklar, Blue, 6 in., Blunt/Blunt, Angled, Serrated, Finger Rings / 1 Large Ring, Stainless Steel / Plastic. Boxes of 12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21100
Econo Multi-Cut Utility Scissors, Sklar, Blue, 6 in., Blunt/Blunt, Angled, Serrated, Finger Rings / 1 Large Ring, Stainless Steel / Plastic. Econo Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are straight and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This item is sold in boxes of 12.
Catalog No. 50-143-7522