Learn More
Thermo Scientific™ Utility Trays for Shaker Platforms Available on GSA/VA Contract for Federal Government customers only.
Use these utility trays with 0.5 in. rubber mats with Thermo Scientific™ MaxQ™ high-performance shakers.
Manufacturer:  Thermo Scientific™ 236026
 View more versions of this product
Catalog No. 236026
  Check Availability
Add to cart
Specifications
Description
Use these Thermo Scientific™ Utility trays for Shaker Platforms with 0.5 in. rubber mats with MaxQ™ High Performance Shakers.
Description & Specifications
Specifications
Utility tray
9x18 in.
MaxQ HP 416, 420, 430, 435, 436, 480, 481
MaxQ™ HP 416, 420, 430, 435, 436, 480, 481 Shaker
22.9 x 45.7cm
Utility Tray