Learn More
Biotix™ uTIP™ Filter Pipette Tips for Universal Pipettes, Standard
Designed with Flexfit™ to lower necessary insertion and ejection force by ≈70% to make secure seal. Biotix™ uTIP™ Filter Pipette Tips for Universal Pipettes are manufactured with low retention X-Resin™ giving tighter CV values.
Supplier: Biotix™ 63300042
Description
- Fits all standard pipettes
- FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
- X-Resin drastically reduces sample retention resulting in tighter CV values
- Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
- Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
- Delta filter technology offers aerosol protection and alerts about potential contamination
Certifications
ISO 9001
Specifications
20 μL | |
Sterile | |
Autoclavable | |
Yes | |
Polypropylene |
Filtered | |
Yellow | |
Rack | |
Biotix uTip |