Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UTP11L Monoclonal Antibody (OTI5F3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527022
Description
UTP11L Monoclonal antibody specifically detects UTP11L in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
UTP11L | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y3A2 | |
UTP11 | |
Full length human recombinant protein of UTP11L produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5F3 | |
Unconjugated | |
UTP11 | |
2700082D03Rik; Cgi94; CGI-94; HDCMB12P; Probable U3 small nucleolar RNA-associated protein 11; RGD1559610; U3 snoRNA-associated protein 11; UTP11; UTP11, small subunit processome component homolog (S. cerevisiae); UTP11L; UTP11-like protein; UTP11-like, U3 small nucleolar ribonucleoprotein; UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast) | |
Mouse | |
Affinity Chromatography | |
RUO | |
51118 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction