Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTP3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549137

Catalog No. PA549137

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Essential for gene silencing: has a role in the structure of silenced chromatin. Plays a role in the developing brain.
Specifications

Specifications

UTP3
Polyclonal
PBS with 0.09% sodium azide
Q9NQZ2
Something about silencing protein 10, Charged amino acid-rich leucine zipper 1, CRL1, Disrupter of silencing SAS10, UTP3 homolog, UTP3, CRLZ1, SAS10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57050
Western Blot
Unconjugated
UTP3
Liquid
UTP3
KLH conjugated synthetic peptide between 416-444 amino acids from the C-terminal region of human UTP3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit