Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTS2D Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17992720UL

 View more versions of this product

Catalog No. NBP17992720


Add to cart

Description

Description

UTS2D Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

UTS2D
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
UTS2D
The immunogen for this antibody is UTS2D. Peptide sequence ALPNKLEELNQLEKLKEQLVEEKDSETSYAVDGLFSSHPSKRACFWKYCV.
13 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
NP_937795
MGC138371, U2B, U-IIB, urotensin 2 domain containing, Urotensin IIB, Urotensin II-related peptide, Urotensin-2 domain-containing protein, urotensin-2B, URPUIIB, UTS2B
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
257313
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit