Learn More
Aviva Systems Biology Corp UTX ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP39483_P050
This is a rabbit polyclonal antibody against UTX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UTX. Alternate names: DKFZp686A03225, MGC141941, bA386N14.2, UTX, KABUK2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53140