Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTY Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$104.00 - $345.50

Specifications

Antigen UTY
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB120178
View Documents
Novus Biologicals
NBP31101525UL
25 μg
Each for $104.00
Add to cart
 
NB120177
View Documents
Novus Biologicals
NBP311015100UL
100 μg
Each for $345.50
Add to cart
 
Description

Description

UTY Polyclonal antibody specifically detects UTY in Human samples. It is validated for Western Blot
Specifications

Specifications

UTY
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
7404
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
DKFZp686L12190, EC 1.14.11, EC 1.14.11.-, histone demethylase UTY, tetratricopeptide repeat protein, ubiquitous TPR motif protein UTY, ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome, ubiquitously transcribed tetratricopeptide repeat gene, Y-linked, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 101, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 106, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 11, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 118, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 121, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 132, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 133, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 136, ubiquitou
The immunogen is a synthetic peptide directed towards the N terminal region of human UTY (NP_009056). Peptide sequence LYETQRKYHSAKEAYEQLLQTENLPAQVKATVLQQLGWMHHNMDLVGDKA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit