Learn More
Cell Biolabs IncSupplier Diversity Partner UV DAMAGE CELL ELISA CPD 96W  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA326
OxiSelect Cellular UV-Induced DNA Damage ELISA Kit measures cyclobutane pyrimidine dimers (CPD) in the DNA of intact cells. 96 wells including standards and unknowns.
Catalog No. 50-673-515