Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ UV Transilluminator 2020L
SDP

Syngene™ UV Transilluminator 2020L is for illumination of any sample excited by UV light.

Supplier:  Syngene™ GE2020LE

 View more versions of this product

Catalog No. 01-257-220

Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.

Alert:

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.


Description

Description

2020L 230V 50Hz 230W 241mm x 315mm x 80mm 364nm wavelength 200mm x 200mm filter area for use with IG3 and UG3 systems Syngene
Specifications

Specifications

364 nm
IG3 and UG3 systems
200 x 200 mm
230 V 50 Hz 230 w
Syngene 2020L
50 Hz
UV Transilluminator
230 w
241 x 315 x 80 mm
200 x 200 mm
230 V
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit