Learn More
Syngene™ UV Transilluminator 2020S
SDP
Syngene™ UV Transilluminator 2020SUV is for illumination of any sample excited by UV light.
Manufacturer:  Syngene™ GE2020SE
 View more versions of this product
Catalog No. 01-257-224
Add to cart
Description
Description
2020S 230V 50Hz 230W 241mm x 315mm x 80mm 254nm wavelength 200mm x 200mm filter area for use with IG3 and UG3 systems Syngene
Specifications
Specifications
254 nm
UV Transilluminator
230 w
241 x 315 x 80 mm
200 x 200 mm
230 V
Fixed
IG3 and UG3 systems
200 x 200 mm
230 V 50 Hz 230 w
Syngene 2020S-E
50 Hz