Learn More
Syngene™ UV Transilluminator 2020S
SDP
Syngene™ UV Transilluminator 2020SUV is for illumination of any sample excited by UV light.
Manufacturer:  Syngene™ GX2020SE
 View more versions of this product
Catalog No. 01-257-252
Add to cart
Description
Description
2020S 230V 50Hz 230W 335mm x 310mm x 100mm 254nm wavelength 200mm x 200mm filter area for use with G:BOX systems Syngene
Specifications
Specifications
254 nm
UV Transilluminator
230 w
335 x 310 x 100 mm
200 x 200 mm
230 V
Fixed
G:Box systems
200 x 200 mm
230 V 50 Hz 230 w
Syngene 2020S-E
50 Hz