Learn More
Honeywell™ Uvex™ Ambient OTG Safety Eyewear Replacement Lens
Designed to fit over most prescription eyewear, and has a lifetime frame guarantee and economical lens replacement system. Honeywell™ Uvex™ Ambient OTG Safety Eyewear offers exceptional peripheral vision and coverage in a lightweight, wrap-around uni-lens design.
$17.81
Specifications
Lens Type IR Shade 5, Scratch-resistant
View More Specs
Products 1
Catalog Number Mfr. No. Lens Tint Lens Type Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Lens Type Price Quantity & Availability  
18-813-423
View Documents
Honeywell™
S6455
Scratch-resistant Each for $17.81
Add to cart
Description
Description
  • The Uvex™ Ambient provides over-the-glass coverage, offering your workers full protection while wearing their own prescription eyewear
  • Available with a range of comfort features and in two sizes, the Uvex™ Ambient OTG offers all-day coverage for a variety of workers and a wide range of prescription eyewear styles
  • This high performance model can improve your workers' productivity as well as lower your cost-of-ownership with Uvex's renowned tints and lens replacement system
  • The Uvex™ Ambient™ OTG is lightweight, comfortable and a stylish alternative to prescription safety eyewear
  • Exceptional peripheral vision and coverage in a lightweight, wraparound uni-lens design
  • Economical and easy lens replacement allows multiple tints and extends the life of the eyewear
Specifications
Specifications
IR Shade 5, Scratch-resistant