Uvex™ Astro OTG 3001™ Safety Glasses
Over-the-glasses style
Manufacturer: Honeywell Safety Products S2534C
 View more versions of this product
Catalog No. 19-084901
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Material Polycarbonate
    Style Over-the-glasses
    Surface Coating UV extreme
    UV Protection 99.90%
    Lens Coating Uvextreme Antifog
    Type Spectacles
    Certifications/Compliance ANSI Z87.1-2003 and CSA Z94.3 standards
    Frame Color Red/White/Blue
    Frame Material Nylon
    Lens Tint Gray XTR
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback