Learn More
Honeywell Safety Products™ Uvex™ Astro OTG 3001™ Safety Glasses Get IT Item
Over-the-glasses style
Manufacturer:  Honeywell Safety Products™ S2510
 View more versions of this product
Catalog No. 18-028
Add to cart
Description
Description
Over-the-glasses style
Order Info
Customization with a company name, logo, or department is available. For details, call your Fisher Scientific Customer Service Representative at 1-800-772-6733.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Specifications
Specifications
Clear UD
Clear
ANSI Z87.1-2015, CSA Z94.3, Military V0 Ballistic Test
Nylon
Polycarbonate
Safety Spectacles; Over-the-glasses
Scratch-resistant
Blue
99.90%
Ultra-dura Hardcoat
Documents
Accessories