Learn More
Honeywell™ Uvex™ Astro™ Rx 3003 Safety Glasses
Manufacturer:  Honeywell™ S2574
 View more versions of this product
Catalog No. 18-079G
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The astro Rx 3003 spectacles feature the styling and fit of astrospec 3000™, plus a bevel groove for prescription lenses and unique lens carrier replacement system.Lightweight, impact-resistant nylon frames are available in black; blue; yellow and black; and red, white, and blue. Propionate browguard and sideshields provide protection against UV absorption, dust, and flying particles.
Order Info
See frame color guide for color selection.
Specifications
Specifications
Red, White and Blue
Nylon
Clear