Learn More
Honeywell™ Uvex™ Astrospec 3000™ CB (Custom Bridge) Safety Glasses
Manufacturer:  Honeywell™ S2200
 View more versions of this product
Catalog No. 18-079A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Custom fit for comfortable protection
Designed specifically for wearers with low-profile nasal area or large-profile cheekbone area, Astrospec 3000 CB spectacles offer features of Astrospec 3000 spectacles plus a customized lens and nose bridge.
Spectacles are lightweight and impact-resistant, with polycarbonate sideshields and browguard for protection against UV, dust, and flying particles. Black frames available with stratch-resistant Ultradura™ coating or Uvextreme™ (XTR) coating for optimal protection from harmful UV light, scratches, fogging, and static.
Order Info
Customization with a company name, logo, or department is available. For details, call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Compliance
Both lens coatings meet ANSI Z87.1+ standards.
Specifications
Specifications
Meets ANSI Z87.1+ standards.
Ultra-dura Hardcoat
Scratch-resistant
Black
Clear