Learn More
Honeywell™ Uvex Avatar™ Safety Glasses
Click to view available options
Frame Color:
Black
Blue
Red
Teal
Lens Tint:
Amber
Clear
Gray
Description
Polycarbonate Lens Coatings available in:
- Hydroshield Anti-fog for 90X longer than the majority of other anti-fog products and 2X the scratch resistance than our next best anti-fog coating
- Hardcoat
Fit:
- Built with MMT—Multi-Material Technology providing a high level of adjustability
- 9-base wrap-around lens provides uncompromised peripheral vision and protection
- Ratchet lens: angle-adjustable temple length and lens inclination of 15 degrees
- Soft, wire-core temples to customize to any facial profile
- Flexible nose pads to fit any nasal profile and reduce slippage
- Soft brow frame with indirect venting
- Frames available in a variety of colors
Protection:
- Certified to meet ANSI+ and tested by Honeywell to meet the requirements of the CSA Z94.3 standard
- “U6” grade -level filtering >99.9% of UVB/UVA protection for far and near UV transmittance
- Available in 4 frame colors and 5 lens tints
Style
- ¾ frame, High performance, contemporary, stylish
Specifications
Certifications/Compliance | ANSI Z.87.1+,CSA Z94.3 |
Lens Type | Hydroshield AF |
Lens Tint | Clear |
Frame Color | Teal |
Temple Style | Bayonette |
Temple Color | Black |
UV Protection | U6 grade |