Learn More
Honeywell™ Uvex™ ExtremePro™ Eyewear
Stands up to the toughest fogging, scratching and sweating
Supplier: Honeywell™ SX0301D
This item is not returnable.
View return policy
Description
- FlexFit wire core temples bend and flex for a customizable, full-comfort fit
- Flexible and replaceable nosepiece conforms to virtually any nasal profile and provides a contoured, non-slip fit
- Replaceable MoistureWick™ brow accessory wicks away moisture to keep lenses clear in high humidity and heat
- GripClick™ frame adjustment lever allows wearer to customize the width of the frame
- Black/gray frame
Used for foundries, chemical plants, metalworking, machining, pulp and paper, shipbuilding or oil and gas industries
Compliance
Exceeds ANSI™ Z87.1-2003.
Specifications
Meets ANSI Z87.1-2003 and certified to CSA Z94.3 standard | |
Scratch-resistant, Anti-fog | |
Minimizes strong outdoor sunlight and glare | |
Moisture-wicking brow | |
Dura-streme | |
Nylon |
Lifetime frame guarantee | |
Espresso | |
Black, Gray | |
Ratcheting | |
Polycarbonate |