Uvex™ ExtremePro™ Eyewear
Stands up to the toughest fogging, scratching and sweating
Manufacturer: Honeywell Safety Products SX0312
 View more versions of this product
Catalog No. 18-813-474
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Warranty Lifetime frame guarantee
    Frame Color Black/Gray
    Lens Coating Supra-Dura Anti-scratch
    Temple Color Black/Gray
    Temple Style Ratcheting
    Lens Type Ultra anti-scratch
    For Use With (Application) Minimizes eye fatigue due to outdoor sunlight and glare
    Certifications/Compliance Meets ANSI Z87.1-2003 and certified to CSA Z94.3 standard
    Frame Material Nylon
    Includes Moisture-wicking brow
    Lens Material Polycarbonate
    Lens Tint Dark Gray
  • Description & Specifications
Related Products
Product Content Feedback