Learn More
Honeywell™ Uvex™ Falcon Safety Eyewear
Comfort and high coverage providing complete and unobstructed vision
Manufacturer:  Honeywell™ S4502X
 View more versions of this product
Catalog No. 19-073-213
Add to cart
Description
Description
  • Sleek, frameless design with wrap-around 9-base unilens
  • Weightless (22g)
  • Lifetime frame guarantee and economical lens replacement system
  • Made in USA
Certifications
Meets ANSI Z87+ (High Impact) and is certified to CSA Z94.3 standards
Specifications
Specifications
Safety
Amber
Spatulite
Black
Anti-fog