Uvex™ Fitlogic™ Safety Glasses
Four unique fitting points adjust to match your exact fit profile
Manufacturer: Honeywell Safety Products S0405X
 View more versions of this product
Catalog No. 19-130-2442
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Warranty Lifetime frame guarantee
    Lens Coating Uvextreme Anti-fog
    Certifications/Compliance Meet ANSI Z87.1-2003 and CSA Z94.3 standards.
    Lens Tint SCT-gray
  • Description & Specifications
Related Products
Product Content Feedback