Learn More
Honeywell™ Uvex™ Flashback™ Safety Glasses
Description
Exclusive single-lens design offers fast, economical lens replacement, but has the look of dual-lens eyewear. Patented Duoflex™ cushions minimize pressure behind the ears, with temple and lens adjustments for a custom fit. Impact-resistant polycarbonate lens and molded-in sideshields and browguard deliver impact protection. Lenses absorb >99.9% of UV radiation. Clear lenses come in your choice of antiscratch Ultradura™ (UD) hardcoat or antifog, antistatic, antiscratch Uvextreme™ AF (XTR) coating, or with gray lenses in UD coating. Black frames.
Compliance
Spectacles meet ANSI Z87.1-2010 standards and are CSA Z94.3-1999 high-impact approved.
Specifications
Certifications/Compliance | Spectacles meet ANSI Z87.1-2003 standards and are CSA Z94.3-1999 high-impact approved. |
Lens Coating | Impact-resistant |
Description | Clear XTR |
Lens Tint | Clear |
Frame Color | Black |
Lens Material | Polycarbonate |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.