Uvex™ Genesis™ Protective Safety Glasses
Three-position ratchet lens inclination system and adjustable temples enable wearer to customize fit
Manufacturer: Honeywell Safety Products S3221X
 View more versions of this product
Catalog No. 19-040-157
  Check Availability
Add to Cart
    Specifications
    Warranty Lifetime frame guarantee
    Frame Color Earth
    Lens Coating Uvextreme Anti-fog
    Certifications/Compliance Meets ANSI Z87+ and CSA Z94.3 standards
    Lens Tint Espresso
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback