Learn More
Uvex™ Genesis™ Protective Safety Glasses
Three-position ratchet lens inclination system and adjustable temples enable wearer to customize fit
Manufacturer:  Honeywell Safety Products S3224
 View more versions of this product
Catalog No. 19-040-160
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lifetime frame guarantee
Scratch-resistant
Earth
SCT-Reflect 50
Meets ANSI Z87+ and CSA Z94.3 standards
Accessories