Uvex™ Genesis™ Protective Safety Glasses
Three-position ratchet lens inclination system and adjustable temples enable wearer to customize fit
Manufacturer: Honeywell Safety Products S3206
 View more versions of this product
Catalog No. 19-003-282
  Check Availability
Add to Cart
    Specifications
    Warranty Lifetime frame guarantee
    Frame Color Black
    Lens Coating Ultra-dura Hardcoat
    Lens Tint Shade 2.0 Infra-dura
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback